missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, Spectrophotometric Grade, 99.5+%, Alfa Aesar

Manufacturer:  Alfa Aesar 32451K2

 View more versions of this product

Catalog No. AA32451K2



Specifications

Specifications

Acetone
Sprectrophotometric
>99.5%
MFCD00008765
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
0.791g/cm3 at 20°C
247 hPa (185mm Hg) at 20°C
1L
67-64-1
Colorless liquid
C3H6O
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(=O)C
58.08
CHEBI:15347
Liquid
Characteristic
Colorless

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(=O)C
Safety and Handling

Safety and Handling

H225-H319-H336: Highly flammable liquid and vapour. Causes serious eye irritation. May cause drowsiness or dizziness.

DOTInformation : DOT Class: 3; Packing Group: II

EINECSNumber : 200-662-2

SDS Documents