Please login to your online account to display your discounted pricing

Acetone (Spectranalyzed™), Fisher Chemical

For use in the ultraviolet and infrared ranges

$142.54 - $302.92

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade Spectranalyzed™
Purity Grade Notes Also meets ACS specifications
Physical Form Liquid
Assay ≥99.5%
Assay Percent Range ≥99.5 %
Molecular Formula C3H6O
Linear Formula CH3COCH3
Formula Weight 58.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

A19-1

  Accessories SDS
fisher scientific company
BPA191
Glass Bottle 1L Each for $142.54
Case of 6 Each for $855.24

A19-4

  Accessories SDS
fisher scientific company
BPA194
Glass Bottle 4L Each for $302.92
Case of 4 Each for $1,211.68
Description & Specifications

Specifications

Chemical Name or Material Acetone
Cleanup Symbol Solvent
Absorbance 0.01 AU max. at 400nm
CAS 67-64-1
ChemAlert Storage Symbol Red
Health Hazard 2 DANGER!
Health Hazard 3 Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0
DOT Information DOT Class 3, : Flammable Liquid
Additional Information Vapor Pressure: 247mbar at 20°C
Aldehyde 0.002% max.
Purity Grade Spectranalyzed™
Purity Grade Notes Also meets ACS specifications
Physical Form Liquid
Assay ≥99.5%
Assay Percent Range ≥99.5 %
Molecular Formula C3H6O
Linear Formula CH3COCH3
Formula Weight 58.08
MDL Number MFCD00008765
UN Number 1090; Colorless
Synonym 2-Propanone
Density 0.790g/cm³
Identification Pass Test
Infrared Spectrum Conforms to reference
Isopropyl Alcohol 0.05% max.
Methanol 0.05% max.
Packaging Glass Bottle
Residue after Evaporation 0.001% max.
Solubility in Water Pass Test
Substances Reducing Permanganate Pass Test
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0006mEq/g max.
Vapor Pressure 247mbar at 20°C
Water 0.5% max.
Flash Point -20°C (-4°C)
Viscosity 0.32 mPa/s at 20°C
Decomposition Information >4°C
Boiling Point 56°C
Color Colorless
Melting Point −95°C
pH 7
Quantity 1L

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Emergency Overview
Flammable liquid and vapor. May cause irritation of respiratory tract. Vapors may cause drowsiness and dizziness. Repeated exposure may cause skin dryness or cracking. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur.

NFPA

Health:1
Flammability:3
Instability:0

DANGER!