Learn More
Acetone, Semiconductor Grade, 99.5%, Thermo Scientific Chemicals
Maximum level of impurities: Color (APHA) 10, Evaporation residue 5ppm, Acidity 0.3μeq/g, Alkalinity 0.5μeq/g, H2O 0.5%, Solubility in H2O P.T., Cl 0.2ppm, PO4 0.1ppm
Supplier: Thermo Scientific Chemicals 019392M1
Description
Acetone, semiconductor grade is suitable for use in electronics and circuit board manufacturing. Its luminescence intensity is dependent upon the solution components and the absorption of UV light induces photolysis and production of radials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetone | |
67-64-1 | |
Colorless liquid | |
56°C | |
-17°C (1°F) | |
Characteristic | |
1.359 | |
C3H6O | |
247 hPa (185mm Hg) at 20°C | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 |
°C to 94°C | |
Colorless | |
0.791 | |
4 x 1 L | |
465°C (869°F) | |
Liquid | |
99.5% | |
MFCD00008765 | |
63580 | |
Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils. | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
Semiconductor |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H225-H319-H336-H373
DOTInformation : Hazard Class: 3, Packing Group: II
EINECSNumber : 200-662-2
RTECSNumber : AL3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only