Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetone semicarbazone, 96%

$37.36 - $206.43

Chemical Identifiers

Chemical Name or Material Acetone semicarbazone
CAS 110-20-3
Molecular Formula C4H9N3O
Formula Weight 115.14
MDL Number MFCD00014785
Beilstein 970110
Merck Index 14,8444
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1243618

  SDS
alfa aesar™
A1243618
50g Each for $37.36

AAA1243622

  SDS
alfa aesar™
A1243622
100g Each for $62.68

AAA1243636

  SDS
alfa aesar™
A1243636
500g Each for $206.43
Description & Specifications

Specifications

Formula Weight 115.14
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Acetone semicarbazone
Health Hazard 1 Danger
RTECS Number AL7175000
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Merck Index 14,8444
CAS 110-20-3
Melting Point 179° to 185°C
TSCA TSCA
Assay 96%
Beilstein 970110
Molecular Formula C4H9N3O
MDL Number MFCD00014785
Quantity 50g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.