missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetone semicarbazone, 96%

Manufacturer:  Alfa Aesar™ A1243636

 View more versions of this product

Catalog No. AAA1243636



Specifications

Specifications

Acetone semicarbazone
500g
C4H9N3O
970110
acetone semicarbazone, acetonesemicarbazone, acetone, semicarbazone, 2-propanone, semicarbazone, hydrazinecarboxamide, 2-1-methylethylidene, isopropylideneamino urea, propan-2-ylideneamino urea, 2-propan-2-ylidene hydrazinecarboxamide, 2-propanone semicarbazone, acmc-1bs9q
CC(=NNC(=O)N)C
115.136
115.14
179°C to 185°C
110-20-3
MFCD00014785
14,8444
HQDAJGNZGNZGCO-UHFFFAOYSA-N
(propan-2-ylideneamino)urea
66965
96%

Chemical Identifiers

110-20-3
115.136
HQDAJGNZGNZGCO-UHFFFAOYSA-N
66965
CC(=NNC(=O)N)C
C4H9N3O
MFCD00014785
acetone semicarbazone, acetonesemicarbazone, acetone, semicarbazone, 2-propanone, semicarbazone, hydrazinecarboxamide, 2-1-methylethylidene, isopropylideneamino urea, propan-2-ylideneamino urea, 2-propan-2-ylidene hydrazinecarboxamide, 2-propanone semicarbazone, acmc-1bs9q
(propan-2-ylideneamino)urea
Safety and Handling

Safety and Handling

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : AL7175000

TSCA : TSCA

SDS Documents