Learn More
Acetone oxime, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 102390025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetone oxime | |
127-06-0 | |
100.0 | |
135.0°C | |
Authentic | |
Plastic Bottle | |
(CH3)2C=NOH | |
2.5kg | |
13,22 | |
acetone oxime, acetoxime, propan-2-one oxime, 2-propanone, oxime, 2-propanone oxime, acetonoxime, acetone, oxime, beta-isonitrosopropane, acetoneoxime, ccris 5 | |
PXAJQJMDEXJWFB-UHFFFAOYSA-N | |
N-propan-2-ylidenehydroxylamine | |
67180 | |
73.09 |
98% | |
97.5 | |
0.9010g/mL | |
60°C | |
97.5% min. (GC) | |
C3H7NO | |
MFCD00002118 | |
01, 649 | |
15, 73 | |
Solubility in water: 330g/L (20°C). Other solubilities: soluble in alcohols and ethers | |
CC(=NO)C | |
73.09 | |
CHEBI:15349 | |
98% |
Safety and Handling
GHS H Statement
Flammable solid.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call
GHS Signal Word: Danger
EINECSNumber : 204-820-1
RTECSNumber : AL6825000
TSCA : TSCA
RUO â Research Use Only