Please login to your online account to display your discounted pricing

Acetone Oxime 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  102395000

 View more versions of this product

Catalog No. AC102395000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 73.09
Physical Form Crystals or Crystalline Solid
Quantity 500g
Color White to yellow
Beilstein 01, 649
Merck Index 15, 73
Fieser 13,22
TSCA TSCA
EINECS Number 204-820-1
RTECS Number AL6825000
Melting Point 58° to 63°C
CAS Max % 100.0
Infrared Spectrum Authentic
Chemical Name or Material Acetone oxime
MDL Number MFCD00002118
Linear Formula (CH3)2C=NOH
Assay Percent Range 98%
Name Note 98%
CAS 127-06-0
Molecular Formula C3H7NO
CAS Min % 97.5
Synonym Acetoxime
Packaging Glass bottle

Specifications

Chemical Name or Material Acetone oxime
Name Note 98%
CAS 127-06-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H7NO
Linear Formula (CH3)2C=NOH
Formula Weight 73.09
MDL Number MFCD00002118
Synonym Acetoxime