missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone Oxime 98%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 102391000

 View more versions of this product

Catalog No. AC102391000


 

Specifications Description & Specifications

Specifications

≥97.5%
73.09
White to yellow
01, 649
58° to 63°C
98%
98%
MFCD00002118
Acetone oxime
Glass bottle
Acetoxime
Crystals or Crystalline Solid
100g
13,22
15, 73
100.0
C3H7NO
97.5
127-06-0
(CH3)2C=NOH
Authentic

Chemical Identifiers

127-06-0
100.0
C3H7NO
73.09
Acetoxime
≥97.5%
97.5
98%
(CH3)2C=NOH
MFCD00002118
Acetone oxime, 98%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 204-820-1

RTECSNumber : AL6825000

TSCA : TSCA