missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetone (OmniSolv™) 99.5%, MilliporeSigma™
Supplier: Technidata AX01166
Specifications
Acetone, 99.5% | |
0.005AU max at 350nm, 0.005AU max at 400nm, 0.06AU max at 340nm, 0.30AU max at 335nm, 0.80AU max at 330nm, 1.00AU max at 329nm | |
1 L | |
Glass Bottle | |
0.3μeq/g max | |
CH3COCH3 | |
0.5% max | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 | |
OmniSolv™ |
67-64-1 | |
0.79kg/L | |
To Pass Test | |
1ppm max | |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.5% (GC, corrected for water content) |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Safety and Handling
Recommended Storage : Room Temperature