missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone (NF), Macron Fine Chemicals™

macron

Manufacturer:  Avantor Performance 243208

 View more versions of this product

Catalog No. MK24324

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Acetone
C3H6O
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
Glass
67-64-1
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(=O)C
58.08
CHEBI:15347
BP/EP/FCC/JP/NF/USP
4L

Chemical Identifiers

67-64-1
58.08
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(=O)C
C3H6O
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
Documents