Please login to your online account to display your discounted pricing

Acetone (NF), Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade BP/EP/FCC/JP/NF/USP
Molecular Formula CH3COCH3
Formula Weight 58.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK243220

 
avantor performance
243219
Steel Pail 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK24324

 
avantor performance
243208
Glass 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade BP/EP/FCC/JP/NF/USP
Formula Weight 58.08
Quantity 20L
CAS 67-64-1
Packaging Steel Pail
Molecular Formula CH3COCH3
Chemical Name or Material Acetone