Please login to your online account to display your discounted pricing

Acetone, Multi-Compendial, N.F., J.T.Baker™

$114.08 - $1,171.10

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade Multi-Compendial/NF
Molecular Formula (CH3)2CO
Formula Weight 58.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002163

 
avantor performance
903601
Narrow Mouth Amber Glass Bottle 500mL Each for $114.08

02002162

 
avantor performance
903603
Narrow Mouth Amber Glass Bottle 4L Each for $1,171.10
Description & Specifications

Specifications

Purity Grade Multi-Compendial/NF
Formula Weight 58.08
Chemical Name or Material Acetone
CAS 67-64-1
Molecular Formula (CH3)2CO
Quantity 500mL
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 0.79kg
Flash Point −20.00°C