Learn More
Acetone, HPLC Grade, 99.5+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 022928K7
Description
Acetone, HPLC Grade is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry. It is useful as a laboratory reagent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetone | |
67-64-1 | |
Colorless liquid | |
56°C | |
-17°C (1°F) | |
Liquid | |
>99.5% | |
MFCD00008765 | |
635680 | |
Miscible with water,benzene,alcohol,dimethylformamide,ether,chloroform and most oils. | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
HPLC |
°C to 94°C | |
Colorless | |
0.791 | |
4 L | |
Characteristic | |
1.359 | |
C3H6O | |
247 hPa (185mm Hg) at 20°C | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Safety and Handling
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P370+P378q-P501c
H225-H319-H336-H373
DOTInformation : DOT Class: 3; Packing Group: II
EINECSNumber : 200-662-2
RTECSNumber : AL3150000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only