Please login to your online account to display your discounted pricing

Acetone, HPLC Grade, 99.5+%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade HPLC Grade
Molecular Formula C3H6O
Linear Formula CH3COCH3
Formula Weight 58.08
MDL Number MFCD00008765
UN Number UN1090
Beilstein 635680
Merck Index 14,66
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA22928M6

  SDS
alfa aesar
22928M6
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 58.08
Purity Grade HPLC Grade
Quantity 4x4L
TSCA TSCA
UN Number UN1090
Density 0.791
Beilstein 635680
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Boiling Point 56°C
CAS 67-64-1
Merck Index 14,66
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONE
Molecular Formula C3H6O
Refractive Index 1.3590
MDL Number MFCD00008765
Health Hazard 2 GHS H Statement
H225-H319-H336;
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Chemical Name or Material Acetone
Color Colorless
Linear Formula CH3COCH3
Health Hazard 1 Danger
Physical Form Liquid
Flash Point −17°C (1°F)
Odor Characteristic
Assay ≥99.5%
RTECS Number AL3150000
Melting Point −94°C
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a;
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H336;
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.