Please login to your online account to display your discounted pricing

Acetone, for gas chromatography ECD and FID SupraSolv™, EMD Millipore

For gas chromatography ECD and FID SupraSolv™

$123.57 - $257.60

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
Molecular Formula CH3COCH3
Formula Weight 58.08
UN Number 1090
Synonym Dimethyl ketone, Propanone, 2-Propanone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

M1000121000

 
emd millipore corporation
1000121000
Glass Bottle 1L Each for $123.57

M1000122500

 
emd millipore corporation
1000122500
Glass Bottle 2.5L Each for $257.60
Description & Specifications

Specifications

Boiling Point 56.2°C
Melting Point −95.4°C
pH 5 to 6 (395g/L at 20°C)
Formula Weight 58.08
Quantity 1L
Chemical Name or Material Acetone
Ignition Point 465°C (869°F)
Odor Fruity
Packaging Glass Bottle
Storage Information 15° to 25°C (59° to 77°F); Keep container tightly closed. Keep container in a cool, well-ventilated place. Keep away from heat and source of ignition. Protected from light.
Vapor Pressure 233 hPa (20°C)
Flash Point <−20°C (<−4°F)
Physical Form Liquid
Color Colorless
Molecular Formula CH3COCH3
CAS 67-64-1
UN Number 1090
Synonym Dimethyl ketone, Propanone, 2-Propanone
Solubility Information Soluble in water at 20°C
Health Hazard 1 H225: Highly flammable liquid and vapour.
Health Hazard 2 H319: Causes serious eye irritation.
Description SupraSolv™ solvents offer the largest specified retention time range, a clear baseline and a minimal signal-to-noise ratio leading to reliable and reproducible analysis results.
Density 0.79g/cm3 at 20°C
Health Hazard 3 H336 May cause drowsiness and dizziness.
Safety and Handling

H225: Highly flammable liquid and vapour.

H336 May cause drowsiness and dizziness.

H319: Causes serious eye irritation.