Please login to your online account to display your discounted pricing

Acetone-d6, for NMR, with 0.03% TMS, 99.9% atom D, ACROS Organics™

$55.30 - $239.70

Chemical Identifiers

Chemical Name or Material Acetone-d6
Name Note 99.9 atom% D
CAS 666-52-4
Purity Grade For NMR
Molecular Formula C3D6O
Linear Formula CD3COCD3
Formula Weight 64.13
MDL Number MFCD00044635
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC351380100

  SDS
thermo fisher scientific
35138-0100
Glass bottle 10mL Each for $55.30

AC351380500

  SDS
thermo fisher scientific
35138-0500
Glass bottle 50mL Each for $239.70
Description & Specifications

Specifications

Formula Weight 64.13
Physical Form Liquid
Quantity 10mL
Color Clear colorless
Beilstein 01, III, 2741
EINECS Number 211-563-9
Purity Grade For NMR
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Chemical Name or Material Acetone-d6
Molecular Formula C3D6O
Linear Formula CD3COCD3
Name Note 99.9 atom% D
CAS 666-52-4
MDL Number MFCD00044635
Water 0.1% max. (Coulometric)
Packaging Glass bottle
Deuterium (D) 99.85% min.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

for NMR

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.