missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone - D6, deuteration degree min. 99.9% for NMR spectroscopy MagniSolv™, MilliporeSigma™

For NMR Spectroscopy

$65.97 - $528.77

Chemical Identifiers

CAS 666-52-4
Molecular Formula C3H6O
Molecular Weight (g/mol) 64.117
InChI Key CSCPPACGZOOCGX-WFGJKAKNSA-N
Synonym acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
PubChem CID 522220
ChEBI CHEBI:78217
IUPAC Name 1,1,1,3,3,3-hexadeuteriopropan-2-one
SMILES CC(=O)C
View More Specs

Products 5
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
M1000210005 View Documents MilliporeSigma
1000210005
10 x 0.5mL Glass ampoule Each for $65.97
 
M1000210009 View Documents MilliporeSigma
1000210009
10 x 0.75mL Glass ampoule Each for $79.20
 
M1000210010 View Documents MilliporeSigma
1000210010
10mL Septum ampoule Each for $96.33
 
M1000210025 View Documents MilliporeSigma
1000210025
25mL Glass bottle Each for $175.04
 
M1000210100 View Documents MilliporeSigma
1000210100
100mL Glass bottle Each for $528.77
 
Specifications

Chemical Identifiers

666-52-4
64.117
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
CHEBI:78217
CC(=O)C
C3H6O
CSCPPACGZOOCGX-WFGJKAKNSA-N
522220
1,1,1,3,3,3-hexadeuteriopropan-2-one

Specifications

465°C (869°F)
Glass ampoule
0.32mPa·s (20°C/68°F)
10 x 0.5mL
99.9% min.
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
CC(=O)C
64.117
CHEBI:78217
Liquid
Characteristic
233 hPa at 20°C (68°F)
Colorless
666-52-4
C3H6O
CSCPPACGZOOCGX-WFGJKAKNSA-N
1,1,1,3,3,3-hexadeuteriopropan-2-one
522220
64.12
Acetone - D6