missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetone-d6, 99.9% (Isotopic)

NMR solvent

Manufacturer:  Alfa Aesar™ 1679750

 View more versions of this product

Catalog No. AA1679750



Specifications

Specifications

Acetone-d6
−17° (1°F)
1.3554
Colorless
5x 10g
666-52-4
MFCD00044635
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
CC(=O)C
64.117
CHEBI:78217
Liquid
0.872g/mL
Acetone-like
56°C
-94°C
Isotopic
C3H6O
UN1090
CSCPPACGZOOCGX-WFGJKAKNSA-N
1,1,1,3,3,3-hexadeuteriopropan-2-one
522220
64.13
99.9%

Chemical Identifiers

666-52-4
64.117
CSCPPACGZOOCGX-WFGJKAKNSA-N
522220
1,1,1,3,3,3-hexadeuteriopropan-2-one
C3H6O
MFCD00044635
acetone-d6, 2h6 acetone, hexadeuteroacetone, acetone d6, perdeuteroacetone, deuterated acetone, unii-b0n19b53h8, cd3 2co, 2-propanone-1,1,1,3,3,3-d6, d6-acetone
CHEBI:78217
CC(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONE

SDS Documents