Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetone-d6, 100% (Isotopic)

Chemical Identifiers

Chemical Name or Material Acetone-d6
CAS 666-52-4
Name Note Isotopic
Formula Weight 64.13
MDL Number MFCD00044635
UN Number UN1090
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA42261KV

 
alfa aesar™
42261KV
10 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA42261SA

 
alfa aesar™
42261SA
2 each CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid In Prescored Ampoules, (0.5Ml/ampoule)
Formula Weight 64.13
Chemical Name or Material Acetone-d6
Density 0.872g/mL
Health Hazard 2 GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Name Note Isotopic
Boiling Point 56°C
Melting Point −94°C
Assay 100%
Quantity 10 each
Color Colorless
Flash Point −17° (1°F)
Refractive Index 1.3554
MDL Number MFCD00044635
CAS 666-52-4
UN Number UN1090
Health Hazard 1 Danger
Odor Acetone-like
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: ACETONE
Safety and Handling

Danger

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H225-H319-H336
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.