Learn More
Acetone cyanohydrin, 99%, stabilized, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 102380250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetone cyanohydrin | |
-19.0°C | |
0.9320g/mL | |
73°C | |
98.5% min. (GC) | |
C4H7NO | |
(CH3)2C(OH)CN | |
03,316 | |
0.932 | |
acetone cyanohydrin, acetone cyanhydrin, 2-methyllactonitrile, 2-hydroxyisobutyronitrile, alpha-hydroxyisobutyronitrile, acetoncyanhydrin, 2-cyano-2-propanol, acetoncianidrina, acetonkyanhydrin, acetoncianhidrinei | |
MWFMGBPGAXYFAR-UHFFFAOYSA-N | |
2-hydroxy-2-methylpropanenitrile | |
Approx. 0.3% H3PO4 | |
4.8 mPa.s (20°C) | |
85.11 | |
Liquid |
75-86-5 | |
Colorless to Yellow | |
82.0°C (23.0 mmHg) | |
Authentic | |
Glass bottle | |
1.3990 to 1.4010 | |
25 g | |
01,5; 08,409; 11,1 | |
15,66 | |
Solubility in water: soluble. | |
CC(C)(C#N)O | |
85.11 | |
6406 | |
CHEBI:15348 | |
99% |
Chemical Identifiers
75-86-5 | |
85.11 | |
acetone cyanohydrin, acetone cyanhydrin, 2-methyllactonitrile, 2-hydroxyisobutyronitrile, alpha-hydroxyisobutyronitrile, acetoncyanhydrin, 2-cyano-2-propanol, acetoncianidrina, acetonkyanhydrin, acetoncianhidrinei | |
CHEBI:15348 | |
CC(C)(C#N)O |
C4H7NO | |
MWFMGBPGAXYFAR-UHFFFAOYSA-N | |
6406 | |
2-hydroxy-2-methylpropanenitrile |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTRE or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 200-909-4
RUO – Research Use Only