missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetone CHROMASOLV™, for HPLC, ≥99.8%, Honeywell Riedel-de Haën™
Acetone's luminesence intensity is dependent upon the solution components.1 The absorption of UV light by acetone results in its photolysis and the production of radials.1
Supplier: Honeywell-Chromasolv 348501L
Alert:
Repeated exposure may cause skin dryness or cracking.
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Acetone | |
for HPLC | |
0.791g/mL at 25°C | |
1 L | |
C3H6O | |
MFCD00008765 | |
635680 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08g/mol | |
CHROMASOLV™ |
-94°C | |
67-64-1 | |
56°C (760 mmHg) | |
n20/D 1.359 | |
CH3COCH3 | |
184mmHg (20°C) | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.8% |
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Safety and Handling
P210-P280-P304 + P340 + P312-P305 + P351 + P338-P337 + P313-P403 + P235
H225-H319-H336
EINECSNumber : 200-662-2
RTECSNumber : AL3150000