Please login to your online account to display your discounted pricing

Acetone (Certified ACS Grade), Fisher BioReagents

$83.34 - $246.07

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade Certified ACS Reagent Grade
Assay ≥99.5%
Molecular Formula C3H6O
Linear Formula CH3COCH3
Formula Weight 58.08
Synonym 2-Propanone
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP2403-4

  SDS
fisher scientific company
BP24034
Amber Glass, EcoSafPak™ 4L Each for $83.34
Case of 4 Each for $333.36

BP2403-20

  SDS
fisher scientific company
BP240320
Steel Pail 20L Each for $246.07
Description & Specifications

Specifications

Assay ≥99.5%
Boiling Point 56°C
Melting Point −95°C
Formula Weight 58.08
Quantity 4L
DOT Information DOT Class 3, : Flammable Liquid
Physical Form Liquid
Viscosity 0.32 mPa/s at 20°C
Purity Grade Certified ACS Reagent Grade
Vapor Pressure 247mbar at 20°C
pH 7
Density 0.790g/cm³
Color Colorless
Isopropyl Alcohol 0.05% max.
Substances Reducing Permanganate Pass Test
Synonym 2-Propanone
Methanol 0.05% max.
Linear Formula CH3COCH3
Recommended Storage RT
CAS 67-64-1
Molecular Formula C3H6O
ChemAlert Storage Symbol Red
Residue after Evaporation 0.001% max.
Identification Pass Test
Boiling Range 0.8°C max.
Infrared Spectrum Conforms to reference
Water 0.5% max.
Solubility in Water Pass Test
Aldehyde 0.002% max.
Packaging Amber Glass, EcoSafPak™
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0006mEq/g max.
Cleanup Symbol Solvent
Chemical Name or Material Acetone

Acetone can be used for biomolecular extraction processes.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

RT