Please login to your online account to display your discounted pricing

Acetone-Alcohol, 1 + 1 Decolorizer Solution, for use in Gram Staining, Ricca Chemical

$81.64 - $203.15

Chemical Identifiers

Chemical Name or Material Acetone-Alcohol
Name Note 1 + 1 Decolorizer Solution
CAS 64-17-5
CAS Min % 49.0
CAS Max % 51.0
CAS 67-64-1
CAS Min % 48.63
CAS Max % 50.61
CAS 64-17-5 (Ethyl Alcohol (Ethanol))
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

200-32

 
ricca chemical company
RI20032
Poly Bottle 1L Each for $81.64

200-1

 
ricca chemical company
R02000004A
Poly Bottle 4L Each for $203.15
Description & Specifications

Specifications

Quantity 1L
Chemical Name or Material Acetone-Alcohol
Name Note 1 + 1 Decolorizer Solution
Packaging Poly Bottle
CAS 64-17-5
CAS Min % 49.0
CAS Max % 51.0
CAS Min % 48.63
CAS Max % 50.61
CAS 64-17-5 (Ethyl Alcohol (Ethanol))
CAS 67-64-1

For use in Gram Staining