missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone-Alcohol, 1:1, Decolorizer, Certified, LabChem™

Manufacturer:  LabChem LC104404

 View more versions of this product

Catalog No. LC104404



Specifications

Specifications

Acetone-Alcohol
1:1, Decolorizer
50,45,2.5,2.5
Passes Test
C3H6O
UN1993
0.792g/mL ±0.008
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
Carbon monoxide, Carbon dioxide May release flammable gases
Liquid
Colorless
4L
67-64-1,64-17-5,67-63-0,67-56-1
0.79
Poly Bottle
CH3COCH3
Soluble in water
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(=O)C
58.08
CHEBI:15347
58.08
Certified

Chemical Identifiers

67-64-1
58.08
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(=O)C
C3H6O
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

Recommended Storage : Room Temperature

Documents
Promotions

Promotions