Please login to your online account to display your discounted pricing

Acetone-Alcohol, 1:1, Decolorizer, Certified, LabChem™


Manufacturer: lab chem  LC104402

 View more versions of this product

Catalog No. LC104402

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Certified Grade
Formula Weight 58.08
CAS 67-56-1
Linear Formula CH3COCH3
Packaging Poly Bottle
CAS Max % 50
Density 0.79
CAS Max % 2.5
Health Hazard 2 GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).
Name Note 1:1, Decolorizer
CAS Max % 45
Recommended Storage Room Temperature
UN Number UN1993
Health Hazard 1 Danger
Identification Passes Test
Physical Form Liquid
Chemical Name or Material Acetone-Alcohol
CAS Max % 2.5
CAS 67-63-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.
CAS 67-64-1
Specific Gravity 0.792g/mL ±0.008
Color Clear, colorless
Decomposition Information Carbon monoxide, Carbon dioxide May release flammable gases
Solubility Soluble in water
CAS 64-17-5

Specifications

Chemical Name or Material Acetone-Alcohol
CAS 67-64-1
CAS Max % 50
CAS 64-17-5
CAS Max % 45
CAS 67-63-0
CAS Max % 2.5
CAS 67-56-1
CAS Max % 2.5
Name Note 1:1, Decolorizer
Purity Grade Certified Grade
Linear Formula CH3COCH3
Formula Weight 58.08
UN Number UN1993
Accessories

Danger

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Keep away from heat, hot surfaces, open flames, sparks.
No smoking.
Keep container tightly closed.
Ground/bond container and receiving equipment.
Use explosion-proof electrical, lighting, ventilating equipment.
Use only non-sparking tools.
Take precautionary measures against static discharge.
Do not breathe mist, spray, vapors.
Use only outdoors or in a well-ventilated area.
Wear eye protection, face protection, protective clothing, protective gloves.
Wash exposed skin thoroughly after handling.
If exposed or concerned: Get medical advice/attention.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Call a poison center/doctor if you feel unwell.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
If eye irritation persists: Get medical advice/attention.
In case of fire: Use dry chemical powder, alcohol-resistant foam, carbon dioxide to extinguish.
Store locked up in a cool, well-ventilated place.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause drowsiness or dizziness.
Suspected of damaging fertility or the unborn child.
Causes damage to organs (central nervous system, optic nerve).

Room Temperature