Please login to your online account to display your discounted pricing

Acetone (ACS AR), Macron Fine Chemicals™


macron

Quantity 950mL
Packaging AR-CAN

Manufacturer: avantor performance  243704

 View more versions of this product

Catalog No. MK2437950

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Purity Grade ACS Reagent Grade
Formula Weight 58.08
Quantity 950mL
Chemical Name or Material Acetone
Packaging AR-CAN
Molecular Formula CH3COCH3
CAS 67-64-1

Specifications

Chemical Name or Material Acetone
CAS 67-64-1
Purity Grade ACS Reagent Grade
Molecular Formula CH3COCH3
Formula Weight 58.08