Please login to your online account to display your discounted pricing

Acetone, 99.8%, for HPLC, ACROS Organics™

$83.44 - $159.58

Chemical Identifiers

Chemical Name or Material Acetone
CAS 67-64-1
CAS Min % 99.75
CAS Max % 100.0
Purity Grade For HPLC
Assay ≥99.8%
Assay Percent Range 99.8%
Molecular Formula C3H6O
Linear Formula CH3COCH3
Formula Weight 58.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC268310010

  Accessories SDS
thermo fisher scientific
268310010
1L Each for $83.44

AC268310025

  Accessories SDS
thermo fisher scientific
268310025
2.5L Each for $159.58
Description & Specifications

Specifications

Chemical Name or Material Acetone
Absorbance (1 cm cell vs HPLC−grade water)
CAS 67-64-1
Absorbance Conditions (1 cm cell vs HPLC-grade water)
CAS Min % 99.75
Health Hazard 1 Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Acidity ≤0.0003meq/g
RTECS Number AL3150000
EINECS Number 200-662-2
Additional Information Vapor Pressure: 247mbar at 20°C
TSCA TSCA
Alkalinity ≤0.0003meq/g
Purity Grade For HPLC
Physical Form Liquid
Assay ≥99.8%
Assay Percent Range 99.8%
Molecular Formula C3H6O
Linear Formula CH3COCH3
Formula Weight 58.08
MDL Number MFCD00008765
UN Number 1090; Colorless
Beilstein 01, IV, 3180
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
Synonym 2-Propanone
Density 0.7900g/mL
Filtered to 0.2 micrometer Yes
Identification Pass Test
Residue after Evaporation ≤0.0002%
Water ≤0.1%
Flash Point -20°C (-4°C)
Decomposition Information >4°C
Boiling Point 56°C
Color Clear
Quantity 1L

For HPLC

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.