missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, 99.6%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 423245000

 View more versions of this product

Catalog No. AC423245000




Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Acetone
67-64-1
100.0
0.7900g/mL
500 mL
EcoSafPak™ Glass Bottle
1.3585
99.6%
0.0003 meq/g max.
0.0006 meq/g max.
MFCD00008765
0.32 mPa.s (20°C)
0.5% max. (K.F.)
02,13; 03,4; 06,9
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
ACS Reagent
-95.0°C
99.5
>4°C
56.0°C
-20°C (-4°C)
Liquid
0.001% max.
0.79
C3H6O
CH3COCH3
Vapor Pressure: 247mbar at 20°C
1090; Colorless
01, 635
15, 65
Solubility in water: soluble. Other solubilities: miscible with alcohol, dimethylformamide,, chloroform, ether and most oils
CC(C)=O
58.08
CHEBI:15347
99.6%

Chemical Identifiers

67-64-1
58.08
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
C3H6O
MFCD00008765
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(C)=O
Safety and Handling

Safety and Handling

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only