missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetone, 99.5% min, ACS, MilliporeSigma™
$106.13 - $13644.00
Chemical Identifiers
CAS | 67-64-1 |
---|---|
Molecular Formula | C3H6O |
Molecular Weight (g/mol) | 58.08 |
MDL Number | MFCD00008765 |
InChI Key | CSCPPACGZOOCGX-UHFFFAOYSA-N |
Synonym | acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone |
PubChem CID | 180 |
ChEBI | CHEBI:15347 |
IUPAC Name | propan-2-one |
SMILES | CC(C)=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
MAX01206
|
Technidata
AX01206 |
500 mL | Glass Bottle |
Each for $106.13
|
|
||||
MAX01209
|
Technidata
AX01209 |
1 L | Poly Bottle |
Each for $153.92
|
|
||||
MAX01208
|
Technidata
AX01208 |
4 L | Glass Bottle |
Each for $279.50
|
|
||||
MAX0120P4
|
Technidata
AX0120P4 |
4 L | Poly-coated Glass Bottle |
Each for $309.37
|
|
||||
MAX01203
|
Technidata
AX01203 |
20 L | Metal Can |
Each for $730.00
|
|
||||
MAX012030
|
Technidata
AX012030 |
200 L | Poly Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
67-64-1 | |
58.08 | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
180 | |
propan-2-one |
C3H6O | |
MFCD00008765 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CHEBI:15347 | |
CC(C)=O |
Specifications
-94.2°C | |
Colorless | |
56.1°C | |
-18.15°C (-0.67°F) Closed cup | |
Pungent | |
Liquid | |
99.5% min. | |
C3H6O | |
CH3COCH3 | |
UN 1090 | |
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone | |
CC(C)=O | |
58.08 | |
CHEBI:15347 | |
≥99.5% (GC, corrected for water content) | |
Acetone, For ACS analysis |
67-64-1 | |
0.79g/cm3 at 20°C | |
500 mL | |
464.85°C (868.7°F) | |
Glass Bottle | |
0.001% max | |
0.0003meq/g max | |
0.0006meq/g max | |
MFCD00008765 | |
0.5% max | |
CSCPPACGZOOCGX-UHFFFAOYSA-N | |
propan-2-one | |
180 | |
58.08 | |
ACS/NF/USP |
Safety and Handling
DOTInformation : DOT Class 3
Recommended Storage : Room Temperature