missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetone, 99.5% min, ACS, EMD Millipore

Manufacturer:  Millipore Sigma AX0120T4

 View more versions of this product

Catalog No. MAX0120T4



Specifications

Specifications

Acetone
67-64-1
ACS/NF/USP
C3H6O
UN 1090
CSCPPACGZOOCGX-UHFFFAOYSA-N
propan-2-one
180
58.08
≥99.5% (GC, corrected for water content)
464.85°C (868.7°F)
0.05% max
Metal Can
To pass test
0.0003meq/g max
0.5% max
-18.15°C (-0.67°F) Closed cup
Colorless
4L
For ACS analysis
(as HCHO) 0.002% max
99.5% min.
CH3COCH3
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CC(=O)C
58.08
CHEBI:15347
Liquid
0.79g/cm3 at 20°C
0.05% max
Pungent
0.001% max
To pass test
0.0006meq/g max
General Chemistry
56.1°C
-94.2°C

Chemical Identifiers

67-64-1
58.08
acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
CHEBI:15347
CC(=O)C
C3H6O
CSCPPACGZOOCGX-UHFFFAOYSA-N
180
propan-2-one
Safety and Handling

Safety and Handling

DOTInformation : DOT Class 3

Recommended Storage : Room Temperature

Documents