Please login to your online account to display your discounted pricing

Acetone, 99.5%, for electronic use, residue free, ACROS Organics™


Quantity 4L
Packaging Glass Bottle

Manufacturer: thermo fisher scientific  327840040

 View more versions of this product

Catalog No. AC327840040

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.8%
Purity Grade For electronic use
Boiling Point 56°C
Physical Form Liquid
Melting Point −95°C
Formula Weight 58.08
Quantity 4L
Color Clear
Packaging Glass Bottle
Flash Point -20°C (-4°C)b
UN Number 1090; Colorless
Decomposition Information >4°C
Density 0.7900g/mL
Beilstein 01, 635
Fieser 02,13; 03,4; 06,9
Merck Index 15, 65
CAS Max % 100.0
Chemical Name or Material Acetone
Additional Information Vapor Pressure: 247mbar at 20°C
Methanol 0.05% max. (GC)
Assay Percent Range 99.5%
Arsenic (As) 10ppb max.
Barium (Ba) 20ppb max.
MDL Number MFCD00008765
Heavy Metals (as Pb) 1ppm max.
Iron (Fe) 50ppb max.
CAS Min % 99.75
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Acetic Acid 0.002% max.
Residue after Evaporation 0.0005% max.
Water 0.2% max. (Coulometric)
Linear Formula CH3COCH3
Chlorine (Cl) 2ppm max.
Copper (Cu) 10ppb max.
Name Note Residue Free
Molecular Formula C3H6O
CAS 67-64-1
Synonym 2-Propanone
Health Hazard 2 GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.
Health Hazard 1 Danger

Specifications

Chemical Name or Material Acetone
Name Note Residue Free
CAS 67-64-1
CAS Min % 99.75
CAS Max % 100.0
Purity Grade For electronic use
Assay ≥99.8%
Assay Percent Range 99.5%
Molecular Formula C3H6O
Linear Formula CH3COCH3
Formula Weight 58.08
MDL Number MFCD00008765
Synonym 2-Propanone

For electronic use

Accessories

Danger

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause drowsiness or dizziness.
Causes serious eye irritation.
Highly flammable liquid and vapor.
Repeated exposure may cause skin dryness or cracking.