Please login to your online account to display your discounted pricing

Acetoin, 96%, may exist as mixture of monomer and dimer, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  411955000

 View more versions of this product

Catalog No. AC411955000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥95%
Physical Form Liquid (Monomer) or Powder or Crystals (Dimer)
Formula Weight 88.11
Color Pale yellow to green-yellow or white to yellow
Quantity 500g
Beilstein 01, 827
Merck Index 15, 63
EINECS Number 208-174-1
TSCA TSCA
Specific Gravity 0.995 to 1.019 (25°C)
RTECS Number EL8790000
CAS Min % 95.0
CAS Max % 100.0
Chemical Name or Material Acetoin
Name Note pract., 93%
Assay Percent Range 96%
Molecular Formula C4H8O2
Packaging Glass bottle
Refractive Index 1.4160 to 1.4220 (20°C, 589nm)
Health Hazard 2 GHS P Statement Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation.
Health Hazard 3 GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Infrared Spectrum Authentic
Linear Formula CH3COCH(OH)CH3
MDL Number MFCD00004521
CAS 513-86-0
Synonym 3-Hydroxy-2-butanone
Storage Information may polymerize during storage
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material Acetoin
Name Note pract., 93%
CAS 513-86-0
CAS Min % 95.0
CAS Max % 100.0
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C4H8O2
Linear Formula CH3COCH(OH)CH3
Formula Weight 88.11
MDL Number MFCD00004521
Synonym 3-Hydroxy-2-butanone
Accessories

GHS Signal Word: Warning

GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Flammable liquid and vapor. Causes skin irritation. Causes serious eye irritation.

SDS & Safety Documents