missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetohydroxamic acid, 98%

$67.08 - $274.50

Chemical Identifiers

CAS 546-88-3
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.067
MDL Number MFCD00009994
InChI Key RRUDCFGSUDOHDG-UHFFFAOYSA-N
Synonym acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure
PubChem CID 1990
ChEBI CHEBI:49029
IUPAC Name N-hydroxyacetamide
SMILES CC(=O)NO
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0156906 View Documents Alfa Aesar™
L0156906
5g Each for $67.08
 
AAL0156914 View Documents Alfa Aesar™
L0156914
25g Each for $274.50
 
Description

Description

Applications
Urease inhibitorAcetohydroxamic acid acts as a drug, as an inhibitor of bacterial and plant urease, which is used for urinary tract infections. It is also used in organic synthesis. It acts as an antiurolithic and antibacterial agent. It is involved in the study of complexation of iron(III) with acetohydroxamic acid as well as in the inhibition study of lansoprazole and omeprazole on Helicobacter pyloni. It plays an important role in the insitu generation of nitrosocarbonylmethane as a Diels-Alder dienophile. In addition to this, it is used in the treatment of kidney stones and urinary tract infections.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in a cool place. Moisture sensitive. Incompatible with strong oxidizing agents, acids and alkalis.
Specifications

Chemical Identifiers

546-88-3
75.067
RRUDCFGSUDOHDG-UHFFFAOYSA-N
1990
N-hydroxyacetamide
C2H5NO2
MFCD00009994
acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure
CHEBI:49029
CC(=O)NO

Specifications

86°C to 90°C
546-88-3
MFCD00009994
14,63
RRUDCFGSUDOHDG-UHFFFAOYSA-N
N-hydroxyacetamide
1990
75.07
Acetohydroxamic acid
5g
C2H5NO2
1739019
acetohydroxamic acid, methylhydroxamic acid, acetylhydroxamic acid, acetic acid, oxime, acetohydroximic acid, lithostat, n-acetylhydroxylamine, acetamide, n-hydroxy, acethydroxamsaure, acethydroxamsaeure
CC(=O)NO
75.067
CHEBI:49029
98%
Safety and Handling

Safety and Handling

GHS H Statement
H360
May damage fertility or the unborn child.

GHS P Statement
P281-P201-P202-P308+P313-P405-P501a
Use personal protective equipment as required.
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
IF exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

RTECSNumber : AK8157000