Learn More
Thermo Scientific Chemicals Acetobromo-alpha-D-glucuronic acid methyl ester, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 436290050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetobromo-α-D-glucuronic acid methyl ester | |
97.5 | |
White to Gray | |
97.5% min. (ELSD) (HPLC) | |
C13H17BrO9 | |
5g | |
GWTNLHGTLIBHHZ-SVNGYHJRSA-N | |
methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate | |
397.17 | |
397.17 | |
98% |
21085-72-3 | |
100.0 | |
80°C to 110°C | |
Authentic | |
Glass bottle | |
2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, acetobromo-alpha-d-glucuronic acid methyl ester, acetobromo-alpha-d-glucuronic acid methyl ester, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, 2,3,4-tri-o-acetyl-alpha-d-glucopyranosyl bromide uronic acid methyl ester, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, acetobromo-alpha-d-glucuronic acid, methyl ester, methyl | |
CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C | |
+185° to +195° (20°C, 589nm) (c=1, ethyl acetate) | |
88785 | |
Crystalline Powder or Solid | |
2% (CaCO3) |
Chemical Identifiers
21085-72-3 | |
397.17 | |
2r,3r,4s,5s,6s-2-bromo-6-methoxycarbonyl tetrahydro-2h-pyran-3,4,5-triyl triacetate, acetobromo-alpha-d-glucuronic acid methyl ester, acetobromo-alpha-d-glucuronic acid methyl ester, bromomethyltriacetyl glucuronate, bromo-2,3,4-tri-o-acetyl-alpha-d-glucopyranuronic acid methyl ester, 2,3,4-tri-o-acetyl-alpha-d-glucopyranosyl bromide uronic acid methyl ester, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, methyl 2s,3s,4s,5r,6r-3,4,5-tris acetyloxy-6-bromooxane-2-carboxylate, zlchem 1315, acetobromo-alpha-d-glucuronic acid, methyl ester, methyl | |
methyl (2S,3S,4S,5R,6R)-3,4,5-triacetyloxy-6-bromooxane-2-carboxylate |
C13H17BrO9 | |
GWTNLHGTLIBHHZ-SVNGYHJRSA-N | |
88785 | |
CC(=O)OC1C(C(OC(C1OC(=O)C)Br)C(=O)OC)OC(=O)C |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
GHS Signal Word: Warning
EINECSNumber : 244-203-4
RUO â Research Use Only