missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetoacetanilide, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 102360050

 View more versions of this product

Catalog No. AC102360050


 

Specifications Description & Specifications

Specifications

≥98.5%
Crystalline Powder
White
12, 518
100.0
99%
MFCD00008780
C10H11NO2
CH3COCH2CONHC6H5
102-01-2
177.2
5g
83° to 86°C
15, 57
Authentic
Acetoacetanilide
99%
AAA
98.5
Glass bottle

Chemical Identifiers

102-01-2
100.0
C10H11NO2
177.2
AAA
≥98.5%
98.5
99%
CH3COCH2CONHC6H5
MFCD00008780
Acetoacetanilide, 99%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 202-996-4

RTECSNumber : AK4200000

TSCA : TSCA