missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic-d3 acid-d, for NMR, 99.5% atom D, ACROS Organics™

$53.75 - $236.02

Chemical Identifiers

CAS 1186-52-3
Molecular Formula C2H4O2
Molecular Weight (g/mol) 64.076
MDL Number MFCD00051051
InChI Key QTBSBXVTEAMEQO-GUEYOVJQSA-N
Synonym 2 h? ethan 2 h oic acid
PubChem CID 2723903
IUPAC Name deuterio 2,2,2-trideuterioacetate
SMILES CC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC166210050 View Documents Thermo Fisher Scientific
166210050
5mL Each for $53.75
 
AC166210250 View Documents Thermo Fisher Scientific
166210250
25mL Each for $236.02
 
Description

Description

For NMR
Specifications

Chemical Identifiers

1186-52-3
64.076
QTBSBXVTEAMEQO-GUEYOVJQSA-N
2723903
CC(=O)O
C2H4O2
MFCD00051051
2 h? ethan 2 h oic acid
deuterio 2,2,2-trideuterioacetate

Specifications

1.1300g/mL
40°C
1.3709
0.1% max. (Coulometric)
Colorless
5mL
C2H4O2
MFCD00051051
2 h? ethan 2 h oic acid
CC(=O)O
64.076
64.08
NMR
99.4% min.
Glass bottle
1.13
115.5°C
15.0°C to 16.0°C
1186-52-3
CD3CO2D
02, III, 203
QTBSBXVTEAMEQO-GUEYOVJQSA-N
deuterio 2,2,2-trideuterioacetate
2723903
Liquid
Acetic-d3 acid-d, 99.5 atom%D
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger