Please login to your online account to display your discounted pricing

Alfa Aesar™ Acetic acid-Trimethylpentane, 60/40 (v/v) soln.

Testing of peroxide

Chemical Identifiers

Chemical Name or Material Acetic acid-Trimethylpentane
MDL Number MFCD00236632
UN Number UN2924
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA41895AE

  SDS
alfa aesar™
41895AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

UN Number UN2924
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S.
TSCA TSCA
Concentration 60/40 (v/v) soln.
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Acetic acid-Trimethylpentane
Quantity 100mL
Physical Form Liquid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H224-H304-H314-H318-H336
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.
MDL Number MFCD00236632
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H224-H304-H314-H318-H336
Extremely flammable liquid and vapor.
May be fatal if swallowed and enters airways.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause drowsiness or dizziness.