missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic acid, sodium salt trihydrate, 99+%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 123240010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Acetic acid, sodium salt trihydrate | |
127-09-3,6131-90-4 | |
100.0 | |
99+% | |
CH3COONa·3H2O | |
02, 96 | |
15, 8709 | |
Solubility in water: 762g/L (20 °C). Other solubilities: 1g/19mL alcohol | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
≥99% | |
1 kg | |
Plastic Bottle |
58.0°C | |
99.0 | |
White | |
C2H3NaO2 | |
MFCD00012459 | |
01,1024; 05,591 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium;acetate | |
517045 | |
136.08 | |
Extra Pure | |
Authentic | |
Adhering Crystals |
Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |
RUO – Research Use Only