missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Acetic acid, sodium salt trihydrate, 99+%, for biochemistry, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC446790050

 View more versions of this product

Catalog No. AC446790050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

0.05 max. (3M) at 260nm
0.001% max
Plastic bottle
0.002% max
5kg
99+%
CH3COONa·3H2O
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium;acetate;trihydrate
23665404
136.08
Biochemistry
Acetic acid, sodium salt trihydrate
0.005% max. (in water)
0.0005% max
58.0°C
6131-90-4
C2H3NaO2·3H2O
15,8709
Solubility in water: 762g/L (20°C). Other solubilities: 1g/19mL alcohol
CC(=O)[O-].O.O.O.[Na+]
136.08
CHEBI:32138
99+%

Chemical Identifiers

6131-90-4
136.08
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]
C2H3NaO2·3H2O
AYRVGWHSXIMRAB-UHFFFAOYSA-M
23665404
sodium;acetate;trihydrate
SDS Documents