Please login to your online account to display your discounted pricing

Acetic acid, sodium salt trihydrate, 99+%, ACS reagent, ACROS Organics™

$25.16 - $127.16

Chemical Identifiers

Chemical Name or Material Acetic acid, sodium salt trihydrate
CAS 6131-90-4
CAS Min % 99.0
CAS Max % 100.0
CAS 127-09-3
Purity Grade ACS Reagent Grade
Assay 99.0 to 101.0%
Assay Percent Range 99+%
Molecular Formula C2H3NaO2·3H2O
Linear Formula CH3COONa·3H2O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC424260010

  SDS
fisher scientific acros
424260010
Plastic bottle 1kg Each for $70.31

AC424260250

  SDS
thermo fisher scientific
424260250
Plastic bottle 25g Each for $25.16

AC424260025

  SDS
thermo fisher scientific
AC424260025
Plastic bottle 2.5kg Each for $127.16
Description & Specifications

Specifications

Color White tranparent
Purity Grade ACS Reagent Grade
Physical Form Crystals
Formula Weight 136.08
Quantity 1kg
Assay 99.0 to 101.0%
pH 7.5 to 9.2 (5% soln. at 25°C)
RTECS Number AJ4580000
Beilstein 02, 96
Fieser 01,1024; 05,591
Merck Index 15, 8709
CAS Min % 99.0
CAS Max % 100.0
Chloride 10ppm max.
CAS 6131-90-4
Insoluble Matter 0.005% max.
Magnesium (Mg) 20ppm max.
Chemical Name or Material Acetic acid, sodium salt trihydrate
Assay Percent Range 99+%
Molecular Formula C2H3NaO2·3H2O
Linear Formula CH3COONa·3H2O
Substances Reducing Permanganate Passes Test
Sulfate 20ppm max.
Calcium (Ca) 50ppm max.
MDL Number MFCD00071557
CAS 127-09-3
Packaging Plastic bottle
Phosphate 5ppm max.
Potassium (K) 50ppm max.
Synonym Sodium acetate trihydrate
Health Hazard 1 HYGROSCOPIC
Heavy Metals (as Pb) 5ppm max.
Infrared Spectrum Authentic
Iron (Fe) 5ppm max.
Safety and Handling

HYGROSCOPIC