Please login to your online account to display your discounted pricing

Acetic acid, sodium salt trihydrate, 99.5%, for analysis, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  217115000

 View more versions of this product

Catalog No. AC217115000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥99.4%
Physical Form Crystals
Formula Weight 136.08
Quantity 500g
Color White
Fieser 01,1024; 05,591
Merck Index 15, 8709
Beilstein 02, 96
RTECS Number AJ4580000
Purity Grade For analysis
pH 7.5 to 9 (5% soln. at 20°C)
Packaging Plastic bottle
Iron (Fe) 5ppm max.
Substances Reducing Permanganate 0.005% max. (as HCOOH)
Calcium (Ca) 10ppm max.
Chlorine (Cl) 10ppm max.
CAS Max % 100.0
CAS 6131-90-4
Chemical Name or Material Acetic acid, sodium salt trihydrate
Linear Formula CH3COONa·3H2O
Name Note p.a.
Assay Percent Range 99.5%
MDL Number MFCD00071557
Molecular Formula C2H3NaO2·3H2O
CAS 127-09-3
CAS Min % 99.4
Heavy Metals (as Pb) 5ppm max.
Sulfate 20ppm max.
Infrared Spectrum Authentic
Potassium (K) 50ppm max.
Magnesium (Mg) 5ppm max.
Insoluble in Water 0.005% max.
Aluminum (Al) 5ppm max.
Phosphate 5ppm max.
Synonym Sodium acetate trihydrate
Health Hazard 1 HYGROSCOPIC

Specifications

Chemical Name or Material Acetic acid, sodium salt trihydrate
Name Note p.a.
CAS 6131-90-4
CAS Min % 99.4
CAS Max % 100.0
CAS 127-09-3
Purity Grade For analysis
Assay ≥99.4%
Assay Percent Range 99.5%
Molecular Formula C2H3NaO2·3H2O
Linear Formula CH3COONa·3H2O
Formula Weight 136.08
MDL Number MFCD00071557
Synonym Sodium acetate trihydrate
Accessories

HYGROSCOPIC

SDS & Safety Documents