Please login to your online account to display your discounted pricing

Acetic acid, sodium salt, 99+%, ACS reagent, anhydrous, ACROS Organics™

$54.07 - $85.22

Chemical Identifiers

Chemical Name or Material Acetic acid, sodium salt
CAS 127-09-3
CAS Min % 99.0
CAS Max % 100.0
Purity Grade ACS Reagent Grade
Assay ≥99.0%
Assay Percent Range 99+%
Molecular Formula C2H3NaO2
Linear Formula CH3CO2Na
Formula Weight 82.03
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC424250025

  SDS
thermo fisher scientific
AC424250025
Plastic Bottle 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC424250050

  SDS
thermo fisher scientific
424250050
Glass bottle 5g Each for $54.07

AC424255000

  SDS
thermo fisher scientific
424255000
Plastic Bottle 500g Each for $85.22
Description & Specifications

Specifications

Assay ≥99.0%
Purity Grade ACS Reagent Grade
Physical Form Fine Crystalline Powder
Formula Weight 82.03
Quantity 2.5kg
Color White
Beilstein 02, 96
Fieser 01,1024; 05,591
Merck Index 15, 8709
Packaging Plastic Bottle
EINECS Number 204-823-8
TSCA TSCA
pH 7.0 to 9.2 (5% soln. at 25°C)
RTECS Number AJ4300010
Insoluble Matter 0.01% max.
Chemical Name or Material Acetic acid, sodium salt
Sulfate 30ppm max.
Assay Percent Range 99+%
Chlorine (Cl) 20ppm max.
Molecular Formula C2H3NaO2
MDL Number MFCD00012459
Synonym Sodium acetate
Iron (Fe) 10ppm max.
Loss on Drying 1.0% max. (120°C)
Calcium (Ca) 50ppm max.
Linear Formula CH3CO2Na
CAS 127-09-3
CAS Min % 99.0
Health Hazard 1 HYGROSCOPIC
CAS Max % 100.0
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Magnesium (Mg) 20ppm max.
Phosphate 10ppm max.
Safety and Handling

HYGROSCOPIC