missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetic acid, Puriss., 99-100%, Honeywell™
Puriss., 99-100%
$169.73 - $2083.55
Chemical Identifiers
| CAS | 64-19-7 |
|---|---|
| Molecular Formula | C2H4O2 |
| Molecular Weight (g/mol) | 60.05 |
| MDL Number | MFCD00036152 |
| InChI Key | QTBSBXVTEAMEQO-UHFFFAOYSA-N |
| Synonym | ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure |
| PubChem CID | 176 |
| ChEBI | CHEBI:15366 |
| IUPAC Name | acetic acid |
| SMILES | CC(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
6000027
|
Honeywell Chemicals
272211L |
1 L |
Each for $169.73
|
|
|||||
|
6000028
|
Honeywell Chemicals
2722125L |
2.5 L |
Each for $299.25
|
|
|||||
|
6000029
|
Honeywell Chemicals
272215L |
5 L |
Each for $599.05
|
|
|||||
|
60045757
|
Honeywell Chemicals
272216X1L |
6 x 1 L |
Case of 6 Each for $885.53
|
|
|||||
|
60045755
|
Honeywell Chemicals
272214X25L |
4 x 2.5 L |
Case of 4 Each for $999.85
|
|
|||||
|
60045756
|
Honeywell Chemicals
272214X5L |
4 x 5 L |
Case of 4 Each for $2,083.55
|
|
|||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
| 64-19-7 | |
| 60.05 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| 176 | |
| acetic acid |
| C2H4O2 | |
| MFCD00036152 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CHEBI:15366 | |
| CC(O)=O |
Specifications
| 64-19-7 | |
| CH3CO2H | |
| 2789 | |
| ethanoic acid, ethylic acid, acetic acid, glacial, methanecarboxylic acid, vinegar acid, glacial, acetasol, acide acetique, essigsaeure | |
| CC(O)=O | |
| 60.05 | |
| CHEBI:15366 | |
| 99 to 100% | |
| 1.049g/mL at 25°C | |
| 16% (92°F), 4% (59°F) | |
| 800°F | |
| Acetic Acid |
| C2H4O2 | |
| MFCD00036152 | |
| 506007 | |
| QTBSBXVTEAMEQO-UHFFFAOYSA-N | |
| acetic acid | |
| 176 | |
| 60.05g/mol | |
| puriss. | |
| 1 L | |
| 40°C (104°F) | |
| 2.07 (vs air) |
Safety and Handling
P210-P260-P280-P303 + P361 + P353-P305 + P351 + P338-P370 + P378
H226-H314
EINECSNumber : 200-580-7
RTECSNumber : AF1225000