Please login to your online account to display your discounted pricing

Acetic acid, potassium salt, 99+%, for molecular biology, ACROS Organics™


Quantity 1kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC397120010

 View more versions of this product

Catalog No. AC397120010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Low Melting Solid
Assay ≥99.0% (on dry substance) (Acidimetry)
Formula Weight 98.14
Quantity 1kg
Color White to light brown
Purity Grade For molecular biology
Merck Index 15, 7726
CAS Min % 99.0
CAS Max % 100.0
Chemical Name or Material Acetic acid, potassium salt
Assay Percent Range 99+%
Molecular Formula C2H3KO2
MDL Number MFCD00012458
CAS 127-08-2
Heavy Metals (as Pb) 4ppm max.
Linear Formula CH3CO2K
Synonym Potassium acetate
Packaging Glass bottle

Specifications

Chemical Name or Material Acetic acid, potassium salt
CAS 127-08-2
CAS Min % 99.0
CAS Max % 100.0
Purity Grade For molecular biology
Assay ≥99.0% (on dry substance) (Acidimetry)
Assay Percent Range 99+%
Molecular Formula C2H3KO2
Linear Formula CH3CO2K
Formula Weight 98.14
MDL Number MFCD00012458
Synonym Potassium acetate