missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic acid, potassium salt, 99+%, for biochemistry, Thermo Scientific™
Supplier: Thermo Fisher Scientific 446820025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Biological bufferSpecifications
Acetic acid, potassium salt | |
292.0°C | |
99+% | |
CH3CO2K | |
15,7726 | |
acetic acid potassium salt, acetic acid, potassium salt, acok, diuretic salt, fema no. 2920, koac, octan draselny czech, potassium acetate, potassium ethanoate, potassiumacetate | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.14 | |
Biochemistry | |
0.002% max |
127-08-2 | |
>250°C | |
C2H3KO2 | |
Plastic bottle | |
2.5kg | |
(5 M in water) Clear colorless solution | |
CC(=O)[O-].[K+] | |
98.14 | |
CHEBI:32029 | |
99+% | |
0.003% max | |
0.001% max |
Chemical Identifiers
127-08-2 | |
98.14 | |
acetic acid potassium salt, acetic acid, potassium salt, acok, diuretic salt, fema no. 2920, koac, octan draselny czech, potassium acetate, potassium ethanoate, potassiumacetate | |
CHEBI:32029 | |
CC(=O)[O-].[K+] |
C2H3KO2 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
517044 | |
potassium;acetate |
Safety and Handling
EINECSNumber : 204-822-2
RUO – Research Use Only