missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Acetic acid, potassium salt, 97%, pure, anhydrous, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 149600025
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Acetic acid, potassium salt | |
96.0 | |
2.5kg | |
C2H3KO2 | |
MFCD00012458 | |
01,906; 06,475 | |
15, 7726 | |
Solubility in water: 2694g/L (25°C). Other solubilities: soluble in alcohol, insoluble in ether | |
CC(=O)[O-].[K+] | |
98.14 | |
CHEBI:32029 | |
97% | |
Authentic |
127-08-2 | |
100.0 | |
97% | |
CH3CO2K | |
02, 96 | |
>250°C | |
potassium acetate, acetic acid, potassium salt, diuretic salt, potassium ethanoate, koac, octan draselny czech, potassiumacetate, acetic acid potassium salt, acok, fema no. 2920 | |
SCVFZCLFOSHCOH-UHFFFAOYSA-M | |
potassium;acetate | |
517044 | |
98.14 | |
Pure | |
Plastic bottle |
Safety and Handling
EINECSNumber : 204-822-2
RUO â Research Use Only