Please login to your online account to display your discounted pricing

Acetic Acid, Glacial USP/FCC/ACS, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Acetic Acid, Glacial
CAS 64-19-7
Purity Grade ACS/FCC/USP
Molecular Formula CH3COOH
Formula Weight 60.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK3121212

 
avantor performance
312146
SAFEMOR 2.5L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Acetic Acid, Glacial
CAS 64-19-7
Purity Grade ACS/FCC/USP
Molecular Formula CH3COOH
Formula Weight 60.05
Packaging SAFEMOR
Quantity 2.5L