Please login to your online account to display your discounted pricing

Acetic acid, glacial, J.T.Baker™

Chemical Identifiers

Chemical Name or Material Acetic acid
CAS 64-19-7
Physical Form Liquid
Molecular Formula CH3COOH
Formula Weight 60.05
Synonym Ethanoic acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

14650389

 
avantor performance
V19314
Glass Container 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MKV19318

 
avantor performance
V19318
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Acetic acid
CAS 64-19-7
Physical Form Liquid
Molecular Formula CH3COOH
Formula Weight 60.05
Synonym Ethanoic acid
Packaging Glass Container
Color Clear colorless
Quantity 500mL