Please login to your online account to display your discounted pricing

Acetic Acid, Glacial (Certified ACS Plus), Fisher BioReagents


Quantity 2.5L
Packaging Glass

Manufacturer: fisher scientific company  BP2401212

 View more versions of this product

Catalog No. BP2401-212

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material Acetic Acid, Glacial
Cleanup Symbol Acid
CAS 64-19-7
ChemAlert Storage Symbol Red
Acetic Anhydride 0.01% max.
Recommended Storage RT
DOT Information DOT Class 8, : Corrosive
Aluminum (Al) 0.3ppm max.
Arsenic and Antimony 0.005ppm max.
Boron (B) 0.2ppm max.
Purity Grade Certified ACS Plus Grade
Calcium (Ca) 0.3ppm max.
Physical Form Liquid
Assay ≥99.7%
Chloride 0.4ppm max.
Molecular Formula C2H4O2
Linear Formula CH3COOH
Formula Weight 60.05
Chromium (Cr) 0.2ppm max.
Synonym Vinegar Acid
Density 1.048g/cm³
Copper (Cu) 0.1ppm max.
Dilution Test Pass Test
Gold (Au) 0.3ppm max.
Heavy Metals (as Pb) 0.5ppm max.
Identification Pass Test
Iron (Fe) 0.2ppm max.
Lead (Pb) 0.3ppm max.
Manganese (Mn) 0.3ppm max.
Magnesium (Mg) 0.3ppm max.
Nickel (Ni) 0.1ppm max.
Packaging Glass
Phosphate 1ppm max.
Potassium (K) 0.3ppm max.
Residue after Evaporation 8ppm max.
Sodium (Na) 0.3ppm max.
Substances Reducing Permanganate Pass Test
Substances Reducing Dichromate Pass Test
Sulfate 0.4ppm max.
Tin (Sn) 0.3ppm max.
Titanium (Ti) 0.3ppm max.
Titratable Base 0.0004mEq/g max.
Vapor Pressure 1.52 kPa at 20°C
Zinc (Zn) 0.3ppm max.
Normality 17.4N
Melting Point 16°C
pH 2.5
Color Colorless
Boiling Point 117°C
Quantity 2.5L

Specifications

Chemical Name or Material Acetic Acid, Glacial
CAS 64-19-7
Purity Grade Certified ACS Plus Grade
Assay ≥99.7%
Molecular Formula C2H4O2
Linear Formula CH3COOH
Formula Weight 60.05
Synonym Vinegar Acid

Acetic Acid, Glacial can be used for several biochemical procedures. Some products are packed in Safe Cote™ which give additional safety for chemical and user both.

Accessories
 

RT

 

SDS & Safety Documents