Please login to your online account to display your discounted pricing

Acetic Acid, Glacial, BAKER INSTRA-ANALYZED™ Plus, J.T.Baker™

Acids for critical trace metal analysis; Quality is assured for up to 64 trace metals tested to very low parts-per-billion (ppb) levels

$161.94 - $2,265.59

Chemical Identifiers

Chemical Name or Material Acetic Acid
CAS 64-19-7
Name Note Glacial
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Molecular Formula CH3COOH
Formula Weight 60.05
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002140

 
avantor performance
937505
Poly Bottle 2.5L Each for $368.80

02002141

 
avantor performance
937503
Poly Bottle 4L Case of 4 Each for $2,265.59

02002142

 
avantor performance
937501
PFA Bottle 500mL Each for $161.94
Description & Specifications

Specifications

Formula Weight 60.05
Chemical Name or Material Acetic Acid
CAS 64-19-7
Name Note Glacial
Quantity 2.5L
Purity Grade BAKER INSTRA-ANALYZED™ Plus
Flash Point 39.00°C
Specific Gravity 1L = 1.04kg
Packaging Poly Bottle
Molecular Formula CH3COOH

    For use in ICP (Inductively Coupled Plasma Spectrometry), Trace Metal Analysis; For Laboratory, Research, or Manufacturing Use